Please login to your online account to display your discounted pricing

Mouse anti-HIV Type 1, Clone: HIV1-24/661, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23433505ML

 View all options for this product


For Research Use Only

Catalog No. NBP23433505


Description & Specifications


Antigen HIV Type 1
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Clone HIV1-24/661
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Virus
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P04585
Host Species Mouse
Immunogen Recombinant HIV-1 Gag p24 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 155348
Test Specificity HIV Type 1
Reconstitution Protein A purified