Please login to your online account to display your discounted pricing

Mouse anti-HIV Type 1, Clone: HIV1-24/661, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23463601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23463601

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Virus
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Format Purified
Conjugate Unlabeled
Immunogen Recombinant HIV-1 Gag p24 protein
Host Species Mouse
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.1mg
Antigen HIV Type 1
Gene Accession No. P04585
Clone HIV1-24/661
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Purification Method Purified
Concentration 1mg/mL
Test Specificity HIV Type 1
Regulatory Status RUO
Gene ID (Entrez) 155348
Isotype IgG1, κ