Please login to your online account to display your discounted pricing

Mouse anti-HIV Type 1, Clone: HIV1-24/661, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23463602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23463602

  • / Each
 

Description & Specifications

Specifications

Antigen HIV Type 1
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Clone HIV1-24/661
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P04585
Host Species Murine
Immunogen Recombinant HIV-1 Gag p24 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 155348
Test Specificity HIV Type 1
Reconstitution Protein A purified
Target Species Virus