Please login to your online account to display your discounted pricing

Mouse anti-HIF-1 alpha, Clone: HIF1A-84, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen HIF-1 alpha
Clone HIF1A-84
Concentration 0.2mg/mL
Immunogen Recombinant human HIF1 alpha protein
Gene Accession No. Q16665
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23424801

 
novus biologicals canada
NBP23424801ML
0.1mL Each for $139.00

NBP23424805

 
novus biologicals canada
NBP23424805ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen HIF-1 alpha
Clone HIF1A-84
Concentration 0.2mg/mL
Immunogen Recombinant human HIF1 alpha protein
Gene Accession No. Q16665
Gene ID (Entrez) 3091
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence