Please login to your online account to display your discounted pricing

Mouse anti-HIF-1 alpha, Clone: HIF1A-84, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23424805ML

 View all options for this product


For Research Use Only

Catalog No. NBP23424805


Description & Specifications


Antigen HIF-1 alpha
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Clone HIF1A-84
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q16665
Host Species Mouse
Immunogen Recombinant human HIF1 alpha protein
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3091
Reconstitution Protein A purified