Please login to your online account to display your discounted pricing

Mouse anti-HIF-1 alpha, Clone: HIF1A-84, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23424801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23424801

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Quantity 0.1mL
Format Purified
Isotype IgG2b, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. Q16665
Concentration 0.2mg/mL
Clone HIF1A-84
Purification Method Purified
Formulation PBS with 0.05% BSA.
Immunogen Recombinant human HIF1 alpha protein
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Gene ID (Entrez) 3091
Regulatory Status RUO
Host Species Mouse
Primary or Secondary Primary
Reconstitution Protein A purified
Cross Reactivity Human
Antigen HIF-1 alpha
Conjugate Unlabeled