Please login to your online account to display your discounted pricing

Mouse anti-HIF-1 alpha, Clone: HIF1A-84, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen HIF-1 alpha
Clone HIF1A-84
Concentration 1mg/mL
Immunogen Recombinant human HIF1 alpha protein
Gene Accession No. Q16665
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23463501

 
novus biologicals canada
NBP23463501MG
0.1mg Each for $384.00

NBP23463502

 
novus biologicals canada
NBP23463502MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen HIF-1 alpha
Clone HIF1A-84
Concentration 1mg/mL
Immunogen Recombinant human HIF1 alpha protein
Gene Accession No. Q16665
Gene ID (Entrez) 3091
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence