Please login to your online account to display your discounted pricing

Mouse anti-HIF-1 alpha, Clone: HIF1A-84, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23463501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23463501

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal
Immunogen Recombinant human HIF1 alpha protein
Host Species Mouse
Gene Accession No. Q16665
Gene ID (Entrez) 3091
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Format Purified
Quantity 0.1mg
Concentration 1mg/mL
Primary or Secondary Primary
Clone HIF1A-84
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen HIF-1 alpha
Reconstitution Protein A purified
Purification Method Purified
Conjugate Unlabeled
Regulatory Status RUO
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG2b, κ