Please login to your online account to display your discounted pricing

Mouse anti-Hepatocyte Specific Antigen, Clone: SPM582, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Hepatocyte Specific Antigen
Clone SPM582
Concentration 0.2mg/mL
Immunogen Extract of a formalin-fixed
Formulation PBS with 0.05% BSA. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23285901

 
novus biologicals canada
NBP23285901ML
0.1mL Each for $139.00

NBP23285910

 
novus biologicals canada
NBP23285910ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Hepatocyte Specific Antigen
Clone SPM582
Concentration 0.2mg/mL
Immunogen Extract of a formalin-fixed
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Canine
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)