Please login to your online account to display your discounted pricing

Mouse anti-Hepatocyte Specific Antigen, Clone: SPM582, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23285910ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23285910

  • / Each
 

Description & Specifications

Specifications

Antigen Hepatocyte Specific Antigen
Format Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 1.0mL
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Immunogen Extract of a formalin-fixed
Isotype IgG1
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Clone SPM582
Concentration 0.2mg/mL
Cross Reactivity Canine
Cross Reactivity Human