Please login to your online account to display your discounted pricing

Mouse anti-Hepatocyte Specific Antigen, Clone: SPM582, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23285901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23285901

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Canine
Cross Reactivity Human
Antigen Hepatocyte Specific Antigen
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Isotype IgG1
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone SPM582
Conjugate Unlabeled
Immunogen Extract of a formalin-fixed
Regulatory Status RUO
Concentration 0.2mg/mL