missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Hepatocyte Specific Antigen, Clone: SPM582, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Hepatocyte Specific Antigen
Clone SPM582
Concentration 1mg/mL
Immunogen Extract of a formalin-fixed
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23481401 Novus Biologicals Canada
NBP23481401MG
0.1mg Each for $384.00
NBP23481402 Novus Biologicals Canada
NBP23481402MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Hepatocyte Specific Antigen
1mg/mL
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM582
Extract of a formalin-fixed
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)