Please login to your online account to display your discounted pricing

Mouse anti-Hepatocyte Specific Antigen, Clone: SPM582, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23481402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23481402

  • / Each
 

Description & Specifications

Specifications

Antigen Hepatocyte Specific Antigen
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone SPM582
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Murine
Immunogen Extract of a formalin-fixed
Isotype IgG1
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Hepatocyte Specific Antigen, also called Hepatocyte Paraffin 1 or Hep Par 1, localizes to the mitochondria of hepatocytes.
Reconstitution Protein A purified
Target Species Canine
Target Species Human