Please login to your online account to display your discounted pricing

Mouse anti-Hepatocyte Specific Antigen, Clone: OCH1E5, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Hepatocyte Specific Antigen
Clone OCH1E5
Concentration 0.2mg/mL
Immunogen Extract of a formalin-fixed
Formulation PBS with 0.05% BSA. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23433601

 
novus biologicals canada
NBP23433601ML
0.1mL Each for $139.00

NBP23433605

 
novus biologicals canada
NBP23433605ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Hepatocyte Specific Antigen
Clone OCH1E5
Concentration 0.2mg/mL
Immunogen Extract of a formalin-fixed
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)