Please login to your online account to display your discounted pricing

Mouse anti-Hepatocyte Specific Antigen, Clone: OCH1E5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23433601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23433601

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Canine
Cross Reactivity Human
Immunogen Extract of a formalin-fixed
Antigen Hepatocyte Specific Antigen
Conjugate Unlabeled
Formulation PBS with 0.05% BSA. Protein G purified.
Regulatory Status RUO
Format Purified
Concentration 0.2mg/mL
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Purification Method Purified
Isotype IgG1
Primary or Secondary Primary
Clone OCH1E5
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.1mL