Please login to your online account to display your discounted pricing

Mouse anti-Hepatocyte Specific Antigen, Clone: OCH1E5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23433605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23433605

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Monoclonal or Polyclonal Monoclonal
Quantity 0.5mL
Clone OCH1E5
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen Hepatocyte Specific Antigen
Purification Method Purified
Concentration 0.2mg/mL
Immunogen Extract of a formalin-fixed
Conjugate Unlabeled
Isotype IgG1
Cross Reactivity Canine
Cross Reactivity Human
Host Species Mouse
Formulation PBS with 0.05% BSA. Protein G purified.
Primary or Secondary Primary
Regulatory Status RUO
Format Purified