Please login to your online account to display your discounted pricing

Mouse anti-Hepatocyte Specific Antigen, Clone: OCH1E5, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23469901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23469901

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Monoclonal or Polyclonal Monoclonal
Format Purified
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Canine
Cross Reactivity Human
Regulatory Status RUO
Purification Method Purified
Immunogen Extract of a formalin-fixed
Clone OCH1E5
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Antigen Hepatocyte Specific Antigen
Concentration 1mg/mL
Reconstitution Protein A purified
Isotype IgG1
Quantity 0.1mg
Conjugate Unlabeled