Please login to your online account to display your discounted pricing

Mouse anti-Hepatocyte Specific Antigen, Clone: HSA98, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Hepatocyte Specific Antigen
Clone HSA98
Concentration 1mg/mL
Immunogen HEP-3B human hepatocellular carcinoma cells
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23464502

 
novus biologicals canada
NBP23464502MG
0.2mg Each for $527.00

NBP23464501

 
novus biologicals canada
NBP23464501MG
0.1mg Each for $384.00
Description & Specifications

Specifications

Antigen Hepatocyte Specific Antigen
Clone HSA98
Concentration 1mg/mL
Immunogen HEP-3B human hepatocellular carcinoma cells
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunocytochemistry
Applications Immunofluorescence