Please login to your online account to display your discounted pricing

Mouse anti-Hepatocyte Specific Antigen, Clone: HSA98, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23464501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23464501

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Antigen Hepatocyte Specific Antigen
Host Species Mouse
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Isotype IgG2b, κ
Clone HSA98
Conjugate Unlabeled
Immunogen HEP-3B human hepatocellular carcinoma cells
Quantity 0.1mg
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Applications Immunocytochemistry
Applications Immunofluorescence
Concentration 1mg/mL