Please login to your online account to display your discounted pricing

Mouse anti-Hepatocyte Specific Antigen, Clone: HSA133, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23464401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23464401

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Cross Reactivity Human
Quantity 0.1mg
Immunogen SK-H1A9-2 human hepatocellular carcinoma cells
Regulatory Status RUO
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Concentration 1mg/mL
Antigen Hepatocyte Specific Antigen
Conjugate Unlabeled
Isotype IgG2b, κ
Reconstitution Protein A purified
Primary or Secondary Primary
Format Purified
Clone HSA133
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.