Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: SPM529, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23436905ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23436905

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen hCG Beta
Gene ID (Entrez) 1082
Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Gene Accession No. P01233
Host Species Mouse
Immunogen Recombinant hCG beta protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This MAb reacts with a protein of 22kDa, identified as beta sub-unit of HCG. It does not cross react with the alpha sub-unit.
Clone SPM529
Concentration 0.2mg/mL
Conjugate Unlabeled
Format Purified
Reconstitution Protein A purified