Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: SPM529, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen hCG Beta
Clone SPM529
Concentration 1mg/mL
Immunogen Recombinant hCG beta protein
Gene Accession No. P01233
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23441601

 
novus biologicals canada
NBP23441601MG
0.1mg Each for $384.00

NBP23441602

 
novus biologicals canada
NBP23441602MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen hCG Beta
Clone SPM529
Concentration 1mg/mL
Immunogen Recombinant hCG beta protein
Gene Accession No. P01233
Gene ID (Entrez) 1082
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot