Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: SPM529, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441602MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441602

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Conjugate Unlabeled
Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.2mg
Isotype IgG1, κ
Regulatory Status RUO
Immunogen Recombinant hCG beta protein
Antigen hCG Beta
Format Purified
Clone SPM529
Host Species Mouse
Test Specificity This MAb reacts with a protein of 22kDa, identified as beta sub-unit of HCG. It does not cross react with the alpha sub-unit.
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene ID (Entrez) 1082
Gene Accession No. P01233
Reconstitution Protein A purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL