Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: SPM105, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434505

  • / Each
 

Description & Specifications

Specifications

Immunogen Recombinant hCG beta protein
Formulation PBS with 0.05% BSA.
Regulatory Status RUO
Gene Accession No. P01233
Gene ID (Entrez) 1082
Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone SPM105
Format Purified
Primary or Secondary Primary
Cross Reactivity Human
Isotype IgG1, κ
Concentration 0.2mg/mL
Host Species Mouse
Antigen hCG Beta
Monoclonal or Polyclonal Monoclonal
Quantity 0.5mL
Reconstitution Protein A purified
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Test Specificity This MAb reacts with a protein of 22kDa, identified as beta sub-unit of HCG. It does not cross react with the alpha sub-unit.