Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: SPM105, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434501

  • / Each
 

Description & Specifications

Specifications

Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen hCG Beta
Formulation PBS with 0.05% BSA.
Gene Accession No. P01233
Host Species Mouse
Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Recombinant hCG beta protein
Isotype IgG1, κ
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1082
Test Specificity This MAb reacts with a protein of 22kDa, identified as beta sub-unit of HCG. It does not cross react with the alpha sub-unit.
Reconstitution Protein A purified
Clone SPM105
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Purification Method Purified
Quantity 0.1mL