Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: SPM105, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439202

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Regulatory Status RUO
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P01233
Host Species Mouse
Gene ID (Entrez) 1082
Quantity 0.2mg
Primary or Secondary Primary
Test Specificity This MAb reacts with a protein of 22kDa, identified as beta sub-unit of HCG. It does not cross react with the alpha sub-unit.
Immunogen Recombinant hCG beta protein
Isotype IgG1, κ
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Antigen hCG Beta
Purification Method Purified
Clone SPM105
Concentration 1mg/mL
Format Purified