Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: HCGb/54+ HCGb/459, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen hCG Beta
Clone HCGb/54+ HCGb/459
Concentration 0.2mg/mL
Immunogen Recombinant hCG beta protein (HCGb/54 and HCGb/459)
Gene Accession No. P01233
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23423805

 
novus biologicals canada
NBP23423805ML
0.5mL Each for $321.00

NBP23423801

 
novus biologicals canada
NBP23423801ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen hCG Beta
Clone HCGb/54+ HCGb/459
Concentration 0.2mg/mL
Immunogen Recombinant hCG beta protein (HCGb/54 and HCGb/459)
Gene Accession No. P01233
Gene ID (Entrez) 1082
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting