Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: HCGb/54+ HCGb/459, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen hCG Beta
Clone HCGb/54+ HCGb/459
Concentration 1mg/mL
Immunogen Recombinant hCG beta protein (HCGb/54 and HCGb/459)
Gene Accession No. P01233
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23463201

 
novus biologicals canada
NBP23463201MG
0.1mg Each for $384.00

NBP23463202

 
novus biologicals canada
NBP23463202MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen hCG Beta
Clone HCGb/54+ HCGb/459
Concentration 1mg/mL
Immunogen Recombinant hCG beta protein (HCGb/54 and HCGb/459)
Gene Accession No. P01233
Gene ID (Entrez) 1082
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting