Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: HCGb/54+ HCGb/459, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23463201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23463201

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen Recombinant hCG beta protein (HCGb/54 and HCGb/459)
Isotype IgG1
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 1082
Clone HCGb/54+ HCGb/459
Antigen hCG Beta
Gene Accession No. P01233
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Concentration 1mg/mL
Conjugate Unlabeled