Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: HCGb/54+ HCGb/459, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23463202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23463202

  • / Each
 

Description & Specifications

Specifications

Antigen hCG Beta
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone HCGb/54+ HCGb/459
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01233
Host Species Murine
Immunogen Recombinant hCG beta protein (HCGb/54 and HCGb/459)
Isotype IgG1
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1082
Reconstitution Protein A purified
Target Species Human