Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: HCGb/459, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen hCG Beta
Clone HCGb/459
Concentration 0.2mg/mL
Immunogen Recombinant hCG beta protein
Gene Accession No. P01233
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23423701

 
novus biologicals canada
NBP23423701ML
0.1mL Each for $139.00

NBP23423705

 
novus biologicals canada
NBP23423705ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen hCG Beta
Clone HCGb/459
Concentration 0.2mg/mL
Immunogen Recombinant hCG beta protein
Gene Accession No. P01233
Gene ID (Entrez) 1082
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting