Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: HCGb/459, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423705

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Recombinant hCG beta protein
Conjugate Unlabeled
Cross Reactivity Human
Antigen hCG Beta
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01233
Host Species Mouse
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1082
Reconstitution Protein A purified
Clone HCGb/459
Isotype IgG1, κ
Purification Method Purified
Concentration 0.2mg/mL