Please login to your online account to display your discounted pricing

Mouse anti-hCG Beta, Clone: HCGb/459, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423701

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1082
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Antigen hCG Beta
Conjugate Unlabeled
Gene Accession No. P01233
Isotype IgG1, κ
Regulatory Status RUO
Reconstitution Protein A purified
Concentration 0.2mg/mL
Cross Reactivity Human
Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Purification Method Purified
Immunogen Recombinant hCG beta protein
Quantity 0.1mL
Clone HCGb/459