Please login to your online account to display your discounted pricing

Mouse anti-hCG alpha, Clone: SPM552, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen hCG alpha
Clone SPM552
Concentration 0.2mg/mL
Immunogen Recombinant hCG alpha protein
Gene Accession No. P01215
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23297101

 
novus biologicals canada
NBP23297101ML
0.1mL Each for $139.00

NBP23297110

 
novus biologicals canada
NBP23297110ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen hCG alpha
Clone SPM552
Concentration 0.2mg/mL
Immunogen Recombinant hCG alpha protein
Gene Accession No. P01215
Gene ID (Entrez) 1081
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting