Please login to your online account to display your discounted pricing

Mouse anti-hCG alpha, Clone: SPM552, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23297110ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23297110

  • / Each
 

Description & Specifications

Specifications

Antigen hCG alpha
Format Purified
Applications ELISA
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Formulation PBS with 0.05% BSA.
Gene Accession No. P01215
Host Species Mouse
Immunogen Recombinant hCG alpha protein
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1081
Reconstitution Protein A purified
Clone SPM552
Conjugate Unlabeled
Cross Reactivity Human
Regulatory Status RUO
Concentration 0.2mg/mL