Please login to your online account to display your discounted pricing

Mouse anti-hCG alpha, Clone: SPM552, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23297101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23297101

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Antigen hCG alpha
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01215
Host Species Mouse
Applications ELISA
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Recombinant hCG alpha protein
Isotype IgG1, κ
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 1081
Clone SPM552
Concentration 0.2mg/mL
Cross Reactivity Human
Purification Method Purified
Reconstitution Protein A purified