missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-hCG alpha, Clone: SPM552, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen hCG alpha
Clone SPM552
Concentration 1mg/mL
Immunogen Recombinant hCG alpha protein
Gene Accession No. P01215
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23479402 Novus Biologicals Canada
NBP23479402MG
0.2mg Each for $527.00
NBP23479401 Novus Biologicals Canada
NBP23479401MG
0.1mg Each for $384.00
Specifications Description & Specifications

Specifications

hCG alpha
1mg/mL
P01215
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM552
Recombinant hCG alpha protein
1081
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Immunohistochemistry (Paraffin),Western Blot