Please login to your online account to display your discounted pricing

Mouse anti-hCG alpha, Clone: SPM552, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23479401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23479401

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Conjugate Unlabeled
Immunogen Recombinant hCG alpha protein
Quantity 0.1mg
Antigen hCG alpha
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Host Species Mouse
Isotype IgG1, κ
Gene ID (Entrez) 1081
Purification Method Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01215
Test Specificity This MAb reacts with a protein of ∽13kDa, identified as alpha sub-unit of HCG.
Format Purified
Primary or Secondary Primary
Concentration 1mg/mL
Clone SPM552
Regulatory Status RUO