Please login to your online account to display your discounted pricing

Mouse anti-hCG alpha, Clone: HCGa/53, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23463001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23463001

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01215
Host Species Mouse
Immunogen Recombinant hCG alpha protein
Isotype IgG1, κ
Purification Method Purified
Gene ID (Entrez) 1081
Reconstitution Protein A purified
Clone HCGa/53
Concentration 1mg/mL
Conjugate Unlabeled
Antigen hCG alpha
Format Purified