Please login to your online account to display your discounted pricing

Mouse anti-HAGHL, Clone: 8.IGSH, Novus Biologicals™

$300.00 - $367.12

Specifications

Antigen HAGHL
Clone 8.IGSH
Immunogen Thiol-modified glutathione conjugated to KLH.
Gene ID (Entrez) 84264
Formulation Liquid. In PBS, pH 7.2 containing 0.1mM PMSF. IgG purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19747901

 
novus biologicals canada
NBP19747901MG
0.1mg Each for $367.12

NBP19747925

 
novus biologicals canada
NBP1974790025MG
0.025mg Each for $300.00
Description & Specifications

Specifications

Antigen HAGHL
Clone 8.IGSH
Immunogen Thiol-modified glutathione conjugated to KLH.
Gene ID (Entrez) 84264
Formulation Liquid. In PBS, pH 7.2 containing 0.1mM PMSF. IgG purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence