Please login to your online account to display your discounted pricing

Mouse anti-HAGHL, Clone: 8.IGSH, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1974790025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19747925

  • / Each
 

Description & Specifications

Specifications

Antigen HAGHL
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Clone 8.IGSH
Conjugate Unlabeled
Format Purified
Formulation Liquid. In PBS, pH 7.2 containing 0.1mM PMSF. IgG purified.
Host Species Murine
Immunogen Thiol-modified glutathione conjugated to KLH.
Isotype IgG1
Purification Method Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 84264
Test Specificity Recognizes mammals Glutathione NEM.
Target Species Human
Target Species Mammalia
Target Species Murine
Target Species Rat