Please login to your online account to display your discounted pricing

Mouse anti-HABP1/C1QBP/GC1q R, Clone: 60.11, Novus Biologicals™

$309.05 - $479.90

Specifications

Antigen HABP1/C1QBP/GC1q R
Clone 60.11
Concentration 0.1mg/mL
Immunogen Recombinant C1qBP corresponding to mature C1qBP (amino acids 74-282)
Gene ID (Entrez) 708
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14961901

 
novus biologicals canada
NBP14961901MG
0.1mg Each for $479.90

NBP14961905

 
novus biologicals canada
NBP149619005MG
0.05mg Each for $309.05
Description & Specifications

Specifications

Antigen HABP1/C1QBP/GC1q R
Clone 60.11
Concentration 0.1mg/mL
Immunogen Recombinant C1qBP corresponding to mature C1qBP (amino acids 74-282)
Gene ID (Entrez) 708
Formulation 1mL (100μg/mL) 0.2μm filtered antibody solution in PBS, containing 0.1% bovine serum albumin.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Golden Syrian Hamster
Cross Reactivity Human
Cross Reactivity Rat
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Applications Immunoassay
Applications Immunoprecipitation
Applications Western Blotting