Please login to your online account to display your discounted pricing

Mouse anti-HABP1/C1QBP/GC1q R, Clone: 60.11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14961901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14961901

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Formulation 1mL (100μg/mL) 0.2μm filtered antibody solution in PBS, containing 0.1% bovine serum albumin.
Applications Flow Cytometry
Applications Functional Assay
Applications Immunoassay
Applications Immunoprecipitation
Applications Western Blotting
Antigen HABP1/C1QBP/GC1q R
Storage Requirements Store at 4°C. Do not freeze.
Conjugate Unlabeled
Host Species Mouse
Cross Reactivity Golden Syrian Hamster
Cross Reactivity Human
Cross Reactivity Rat
Clone 60.11
Concentration 0.1mg/mL
Isotype IgG1
Test Specificity The monoclonal antibody 60.11 is directed against epitopes situated within the NH2-terminal stretch of C1qBP corresponding to residues 76-93. Clone 60.11 recognizes the putative C1q binding site and reacts with the mature form, but has poor or no reactivity with the truncated form, lacking residues 74-95.
Immunogen Recombinant C1qBP corresponding to mature C1qBP (amino acids 74-282)
Format Affinity Purified
Regulatory Status RUO
Gene ID (Entrez) 708
Purification Method Affinity Purified