Please login to your online account to display your discounted pricing

Mouse anti-GRP75/HSPA9B/Mortalin, Clone: 30A5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197448005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19744805

  • / Each
 

Description & Specifications

Specifications

Antigen GRP75/HSPA9B/Mortalin
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone 30A5
Conjugate Unlabeled
Format Purified
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Ammonium sulfate precipitation.
Host Species Murine
Immunogen Native human Grp75 (Mortalin).
Isotype IgG1
Purification Method Precipitation
Quantity 0.05mL
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3313
Target Species Bovine
Target Species Canine
Target Species Chicken
Target Species Drosophila
Target Species Guinea Pig
Target Species Hamster
Target Species Human
Target Species Murine
Target Species Porcine
Target Species Primate
Target Species Rabbit
Target Species Rat
Target Species Sheep
Target Species Xenopus