Please login to your online account to display your discounted pricing

Mouse anti-Granulocyte Marker, Clone: SPM250, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Granulocyte Marker
Clone SPM250
Concentration 0.2mg/mL
Immunogen Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes.
Formulation PBS with 0.05% BSA. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23285601

 
novus biologicals canada
NBP23285601ML
0.1mL Each for $139.00

NBP23285610

 
novus biologicals canada
NBP23285610ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Granulocyte Marker
Clone SPM250
Concentration 0.2mg/mL
Immunogen Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes.
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Macaque
Cross Reactivity Monkey
Cross Reactivity Primate
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)