Please login to your online account to display your discounted pricing

Mouse anti-Granulocyte Marker, Clone: SPM250, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23285601ML

 View all options for this product


For Research Use Only

Catalog No. NBP23285601


Description & Specifications


Antigen Granulocyte Marker
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone SPM250
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Macaque
Cross Reactivity Monkey
Cross Reactivity Primate
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Immunogen Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes.
Isotype IgG1
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal