Please login to your online account to display your discounted pricing

Mouse anti-Granulocyte Marker, Clone: SPM250, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475002

  • / Each
 

Description & Specifications

Specifications

Antigen Granulocyte Marker
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone SPM250
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Murine
Immunogen Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes.
Isotype IgG1
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Recognizes an unidentified antigen in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell type in human tissues.
Reconstitution Protein A purified
Target Species Human
Target Species Macaque
Target Species Monkey
Target Species Primate