Please login to your online account to display your discounted pricing

Mouse anti-Granulocyte Marker, Clone: SPM250, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475001

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Macaque
Cross Reactivity Monkey
Cross Reactivity Primate
Antigen Granulocyte Marker
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes.
Isotype IgG1
Purification Method Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Recognizes an unidentified antigen in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell type in human tissues.
Clone SPM250
Conjugate Unlabeled
Concentration 1mg/mL
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified