Please login to your online account to display your discounted pricing

Mouse anti-Golgi Complex, Clone: SPM581, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23285310ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23285310

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Primary or Secondary Primary
Cross Reactivity Human
Antigen Golgi Complex
Formulation PBS with 0.05% BSA.
Immunogen SU-DHL-1 large cell lymphoma cells.
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone SPM581
Concentration 0.2mg/mL
Conjugate Unlabeled