Please login to your online account to display your discounted pricing

Mouse anti-Golgi Complex, Clone: SPM581, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23285301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23285301

  • / Each
 

Description & Specifications

Specifications

Antigen Golgi Complex
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Clone SPM581
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen SU-DHL-1 large cell lymphoma cells.
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified